missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 1.00 N, Ricca Chemical

Catalog No. 6421
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 L
4 L
500 mL
Packaging:
Natural Poly Bottle
Catalog No. Quantity Packaging
6421 4 L Natural Poly Bottle
64216 500 mL Natural Poly Bottle
64232 1 L Natural Poly Bottle
3 options

Catalog No. 6421

Supplier: Ricca Chemical Company RI6421

Only null left

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name amine hydrate
SMILES N.O

Specifications

Color Colorless
CAS Min % 92.0, 5.73
CAS Max % 96.0, 5.96
Concentration or Composition Notes 1.00Normal
Quantity 4 L
Grade Laboratory
Packaging Natural Poly Bottle
Physical Form Liquid
Chemical Name or Material Ammonium Hydroxide