missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Hydroxide, 0.500 N (N/2), Ricca Chemical

Catalog No. 64132
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
64132 1 L Natural Poly Bottle
64116 500 mL Natural Poly Bottle
6411 4 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 64132 Supplier Ricca Chemical Company Supplier No. 64132

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1336-21-6 , 7732-18-5
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
Synonym ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name ammonium hydroxide
SMILES [NH4+].[OH-]

Specifications

Concentration or Composition Notes 0.500Normal
Quantity 1 L
Packaging Natural Poly Bottle
Chemical Name or Material Ammonium Hydroxide