missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorotitanate(IV), 99.99%, (trace metal basis)

Catalog No. p-3777093
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC194350100 10 g Glass bottle
AC194350500 50 g Glass bottle
AC194352500 250 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC194350100 Supplier Thermo Scientific Chemicals Supplier No. 194350100
Only null left

CAS: 16962-40-6 | H8F6N2Ti | 197.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis

Chemical Identifiers

CAS 16962-40-6
Molecular Formula H8F6N2Ti
Molecular Weight (g/mol) 197.97
MDL Number MFCD00010888

Specifications

Color White
Physical Form Crystalline Powder, Crystals and/or Chunks
Quantity 10 g
Assay Percent Range 99.99%
Linear Formula (NH4)2TiF6
Solubility Information Solubility in water: insoluble
Formula Weight 197.97
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ammonium hexafluorotitanate(IV)
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if
EINECSNumber 241-036-9

RUO – Research Use Only