missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorotitanate, 99.99% (metals basis)

Catalog No. AA4319214
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4319214 25 g
AA4319222 100 g
AA4319236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4319214 Supplier Thermo Scientific Chemicals Supplier No. 04319214
Only null left

CAS: 16962-40-6 | (NH4)2TiF6

Ammonium hexafluorotitanate is used as an anti-corrosive cleaning agent. It is also used for the production of ceramics and glass. Further, it is used in the preparation of synthetic gems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16962-40-6
Molecular Formula (NH4)2TiF6
MDL Number MFCD00010888

Specifications

Color White
Physical Form Crystalline
Quantity 25 g
Solubility Information Insoluble in water.
Formula Weight 197.95
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Ammonium hexafluorotitanate
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H302-H312-H332
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 241-036-9
RTECSNumber XR1500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only