missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorophosphate, 99%, pure

CAS: 16941-11-0 | H4F6NP | 163 g/mol

Supplier:  Thermo Scientific Chemicals 202325000

 View more versions of this product

Catalog No. AC202325000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ammonium hexafluorophosphate
99.5%
White to Beige
500 g
H4F6NP
MFCD00064642
ammonium hexafluorophosphate, ammonium phosphorus hexafluoride, ammonium hexafluophosphate, ammonium hexafluophosphate, unii-ns308031pj, unii-ns308031pj, nh4pf6, ksc492g2j, ksc492g2j, e5-phosphanuide, e5-phosphanuide
NIZXKAYXSNUDOU-UHFFFAOYSA-O
azanium;hexafluorophosphate
9793912
99%
Plastic bottle
2.1800g/mL
198°C
16941-11-0
Crystalline Powder or Needles
99%
NH4PF6
15, 523
Solubility in water: 74.8g/100mL (20 c). Other solubilities: soluble in acetone,methanol,ethanol and methyl,acetate
[NH4+].F[P-](F)(F)(F)(F)F
163
163
Pure
2.18

Chemical Identifiers

16941-11-0
163
NIZXKAYXSNUDOU-UHFFFAOYSA-O
9793912
[NH4+].F[P-](F)(F)(F)(F)F
H4F6NP
MFCD00064642
ammonium hexafluorophosphate, ammonium phosphorus hexafluoride, ammonium hexafluophosphate, ammonium hexafluophosphate, unii-ns308031pj, unii-ns308031pj, nh4pf6, ksc492g2j, ksc492g2j, e5-phosphanuide, e5-phosphanuide
azanium;hexafluorophosphate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 241-009-1

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only