missing translation for 'onlineSavingsMsg'
Learn More

Ammonium hexafluorophosphate, 99%, pure

Catalog No. p-222618
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC202320050 5 g Plastic bottle
AC202320250 25 g Plastic bottle
AC202321000 100 g Plastic bottle
AC202325000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC202320050 Supplier Thermo Scientific Chemicals Supplier No. 202320050
Only null left

CAS: 16941-11-0 | H4F6NP | 163 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 16941-11-0
Molecular Formula H4F6NP
Molecular Weight (g/mol) 163
MDL Number MFCD00064642
InChI Key NIZXKAYXSNUDOU-UHFFFAOYSA-O
Synonym ammonium hexafluorophosphate, ammonium hexafluorophosphate v, ammonium hexafluophosphate, ammoniumhexafluorophosphate 6ci,7ci, unii-ns308031pj, azanium hexafluorophosphate, nh4pf6, ksc492g2j, ammonium hexafluoro-, e5-phosphanuide, ammonium phosphorus hexafluoride
PubChem CID 9793912
IUPAC Name azanium;hexafluorophosphate
SMILES [NH4+].F[P-](F)(F)(F)(F)F

Specifications

Melting Point 198°C
Color White to Beige
Physical Form Crystalline Powder or Needles
Quantity 5 g
Assay Percent Range 99%
Linear Formula NH4PF6
Merck Index 15, 523
Solubility Information Solubility in water: 74.8g/100mL (20 c). Other solubilities: soluble in acetone,methanol,ethanol and methyl,acetate
Formula Weight 163
Percent Purity 99%
Grade Pure
Packaging Plastic bottle
Specific Gravity 2.18
Density 2.1800g/mL
Chemical Name or Material Ammonium hexafluorophosphate, 99.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-009-1
TSCA TSCA

RUO – Research Use Only