missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Fluoride, MilliporeSigma™

Catalog No. p-5547609
Change view
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1011640250 250 g Plastic Bottle
M1011641000 1 kg Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1011640250 Supplier Technidata Supplier No. 1.01164.0250
Only null left

For analysis EMSURE™ ACS

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
Synonym Fluoroammonium
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Quantity 250 g
UN Number UN2505
Solubility Information Soluble in water: 820g/L at 20°C
Formula Weight 37.04
Odor Ammoniacal
Packaging Plastic Bottle
Density 1.01g/cm3 at 20°C
Chemical Name or Material Ammonium fluoride
Hazard Category H301: Toxic if swallowed
Hazard Statement H311: Toxic in contact with skin
Precautionary Statement 331: Toxic if inhaled
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 235-185-9
Recommended Storage Store at 15° to 25°C (59° to 77°F), Tightly closed. Dry. Keep in well-ventilated place. Keep locked up or in an area accessible only to qualified or authorized persons.