missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Fluoride (Crystalline/Certified ACS), Fisher Chemical™

Catalog No. A665100
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A665100 100 g Poly Bottle
A665500 500 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A665100 Supplier Thermo Fisher Scientific Supplier No. FLA665100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Color White
pH 6
Physical Form Powder/Solid
Quantity 100 g
Assay Percent Range ≥98.0%
Formula Weight 37.04
Percent Purity ≥98.0%
Grade Certified ACS
Identification Pass Test
Packaging Poly Bottle
Chemical Name or Material Ammonium Fluoride
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation DOT Class 6.1, : Poison