missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, ACS, 98.0% min

CAS: 12125-01-8 | FH4N | 37.04 g/mol

Supplier:  Thermo Scientific Chemicals 011568A1

 View more versions of this product

Catalog No. AA11568A1


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

Ammonium fluoride
12125-01-8
Crystalline
98% min.
MFCD00011423
LDDQLRUQCUTJBB-UHFFFAOYSA-N
amine hydrofluoride
37.04
Hygroscopic
1.01
For analysis ACS
White
1 kg
FH4N
Freely soluble in water. Slightly soluble in alcohol.
N.F
37.04
ACS
White crystalline

Chemical Identifiers

12125-01-8
37.04
LDDQLRUQCUTJBB-UHFFFAOYSA-N
N.F
FH4N
MFCD00011423
amine hydrofluoride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c

H301+H311+H331

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 235-185-9

RTECSNumber : BQ6300000

TSCA : Yes

storageNote1 : Hygroscopic

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only