missing translation for 'onlineSavingsMsg'
Learn More

Ammonium fluoride, ACS, 98.0% min

Catalog No. AA1156814
Change view
Click to view available options
Quantity:
25 g
250 g
1 kg
5 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1156814 25 g
AA1156830 250 g
AA11568A1 1 kg
AA11568D9 5 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1156814 Supplier Thermo Scientific Chemicals Supplier No. 01156814
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 12125-01-8 | FH4N | 37.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Chemical Identifiers

CAS 12125-01-8
Molecular Formula FH4N
Molecular Weight (g/mol) 37.04
MDL Number MFCD00011423
InChI Key LDDQLRUQCUTJBB-UHFFFAOYSA-N
IUPAC Name amine hydrofluoride
SMILES N.F

Specifications

Color White
Physical Form Crystalline
Quantity 25 g
Assay Percent Range 98.0%
Solubility Information Freely soluble in water. Slightly soluble in alcohol.
Formula Weight 37.04
Grade ACS
Sensitivity Hygroscopic
Density 1.01 g/mL
Chemical Name or Material Ammonium fluoride, For analysis ACS
Hazard Category H301+H311+H331
Precautionary Statement P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 235-185-9
RTECSNumber BQ6300000
TSCA Yes
storageNote1 Hygroscopic
Recommended Storage Ambient temperatures

RUO – Research Use Only