missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Dihydrogen Phosphate, MilliporeSigma™
For analysis EMSURE™ ACS, Reag. Ph Eur
Supplier: Technidata 1011260500
Specifications
| Ammonium dihydrogen phosphate | |
| 500 g | |
| MFCD00003396 | |
| Soluble in water: 368g/L at 20°C | |
| N.OP(O)(O)=O | |
| 115.03 | |
| CHEBI:62982 | |
| Odorless | |
| 0.066hPa at 125°C |
| 7722-76-1 | |
| H6NO4P | |
| ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
| LFVGISIMTYGQHF-UHFFFAOYSA-N | |
| phosphoric acid amine | |
| 24402 | |
| 115.02 | |
| Plastic Bottle | |
| 1.80g/cm3 at 20°C |
Chemical Identifiers
| 7722-76-1 | |
| 115.03 | |
| LFVGISIMTYGQHF-UHFFFAOYSA-N | |
| 24402 | |
| phosphoric acid amine |
| H6NO4P | |
| MFCD00003396 | |
| ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate | |
| CHEBI:62982 | |
| N.OP(O)(O)=O |
Safety and Handling
EINECSNumber : 231-764-5
Recommended Storage : Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry