missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Dihydrogen Phosphate, MilliporeSigma™

Catalog No. p-5547608
Change view
Click to view available options
Quantity:
50 g
500 g
Packaging:
Fibre carton
Plastic Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1011269050 50 g Fibre carton
M1011260500 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1011269050 Supplier Technidata Supplier No. 1.01126.9050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For analysis EMSURE™ ACS, Reag. Ph Eur

Chemical Identifiers

CAS 7722-76-1
Molecular Formula H6NO4P
Molecular Weight (g/mol) 115.03
MDL Number MFCD00003396
InChI Key LFVGISIMTYGQHF-UHFFFAOYSA-N
Synonym ammonium dihydrogen phosphate, monoammonium phosphate, ammonium biphosphate, ammonium acid phosphate, phosphoric acid, monoammonium salt, ammonium monophosphate, ammonium dihydrogen orthophosphate, ammonium dihydrophosphate, ammonium diacid phosphate, primary ammonium phosphate
PubChem CID 24402
ChEBI CHEBI:62982
IUPAC Name phosphoric acid amine
SMILES N.OP(O)(O)=O

Specifications

Quantity 50 g
Solubility Information Soluble in water: 368g/L at 20°C
Formula Weight 115.02
Odor Odorless
Packaging Fibre carton
Vapor Pressure 0.066hPa at 125°C
Density 1.80g/cm3 at 20°C
Chemical Name or Material Ammonium dihydrogen phosphate
EINECSNumber 231-764-5
Recommended Storage Store at 5° to 30°C (41° to 86°F), Tightly closed. Dry