missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Dichromate, Crystal, 99.5%, Spectrum™ Chemical
$237.50 - $16326.82
Chemical Identifiers
| CAS | 7789-09-5 |
|---|---|
| Molecular Formula | Cr2H8N2O7 |
| Molecular Weight (g/mol) | 252.06 |
| InChI Key | JOSWYUNQBRPBDN-UHFFFAOYSA-P |
| IUPAC Name | diammonium [(oxidodioxochromio)oxy]chromiumoylolate |
| SMILES | [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18608631
|
Spectrum Chemical Mfg Cor
A117825GM |
25 g | Poly Bottle |
Each for $237.50
|
|
||||
|
18608626
|
Spectrum Chemical Mfg Cor
A1178125GM |
125 g | Poly Bottle |
Each for $445.93
|
|
||||
|
18608628
|
Spectrum Chemical Mfg Cor
A1178500GM |
500 g | Poly Bottle |
Each for $1,085.46
|
|
||||
|
18608630
|
Spectrum Chemical Mfg Cor
A117825KG |
2.5 kg | Poly Pail |
Each for $3,618.32
|
|
||||
|
18608627
|
Spectrum Chemical Mfg Cor
A117812KG |
12 kg | Poly Pail |
Each for $8,115.11
|
|
||||
|
18608629
|
Spectrum Chemical Mfg Cor
A117850KGBL |
50 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 7789-09-5 | |
| 252.06 | |
| diammonium [(oxidodioxochromio)oxy]chromiumoylolate |
| Cr2H8N2O7 | |
| JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
| [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O |
Specifications
| 7789-09-5 | |
| 3 to 5 | |
| Cr2H8N2O7 | |
| [NH4+].[NH4+].[O-][Cr](=O)(=O)O[Cr]([O-])(=O)=O | |
| 252.06 | |
| Reagent |
| 1 | |
| 25 g | |
| JOSWYUNQBRPBDN-UHFFFAOYSA-P | |
| diammonium [(oxidodioxochromio)oxy]chromiumoylolate | |
| 99.5% | |
| Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.