missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Citrate Dibasic, Honeywell Fluka™
$356.37 - $356.37
Chemical Identifiers
| CAS | 3012-65-5 |
|---|---|
| Molecular Formula | C6H14N2O7 |
| Molecular Weight (g/mol) | 226.19 |
| MDL Number | MFCD00013068 |
| InChI Key | YXVFQADLFFNVDS-UHFFFAOYSA-N |
| Synonym | ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss |
| PubChem CID | 13710713 |
| ChEBI | CHEBI:63076 |
| SMILES | [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O |
Chemical Identifiers
| 3012-65-5 | |
| 226.19 | |
| YXVFQADLFFNVDS-UHFFFAOYSA-N | |
| 13710713 | |
| [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O |
| C6H14N2O7 | |
| MFCD00013068 | |
| ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss | |
| CHEBI:63076 |
Specifications
| 3012-65-5 | |
| C6H14N2O7 | |
| MFCD00013068 | |
| ammonium citrate dibasic, ammonium hydrogencitrate, diammonium hydrogen citrate, citric acid diammonium salt, citric acid ammonium salt, di-ammonium hydrogen citrate, citro diamine, citric acid diamine, ammonium citrate, dibasic, acs, ammonium citrate dibasic, puriss | |
| [NH4+].[NH4+].OC(=O)C(O)(CC([O-])=O)CC([O-])=O | |
| 226.19 | |
| CHEBI:63076 | |
| ≥99% (T) | |
| ≤0.01% (as SO4) | |
| Ammonium Citrate Dibasic |
| 500 g | |
| HOC(CO2H)(CH2CO2NH4)2 | |
| 4925760 | |
| YXVFQADLFFNVDS-UHFFFAOYSA-N | |
| azane;2-hydroxypropane-1,2,3-tricarboxylic acid | |
| 13710713 | |
| 226.18g/mol | |
| ACS/puriss. p.a. | |
| Vapor Density: 1.8 (vs air) |
Safety and Handling
P261-P305 + P351 + P338
H319-H335
EINECSNumber : 221-146-3
RTECSNumber : GE7545000