missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Citrate Dibasic, ACS, 98-103%, Spectrum™ Chemical

Catalog No. p-7164985
Change view
Click to view available options
Quantity:
12 kg
125 g
2.5 kg
45 kg
500 g
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Catalog No. Quantity Packaging
18608622 125 g Poly Bottle
18608625 500 g Poly Bottle
18608621 2.5 kg Poly Bottle
18608623 12 kg Poly Pail
18608624 45 kg Fiber Drum
5 options

Catalog No. 18608622

Supplier: Spectrum Chemical Mfg Cor A1175125GM

Only null left

Chemical Identifiers

CAS 3012-65-5

Specifications

CAS Max % 1
Quantity 125 g
Percent Purity 98 to 103%
Grade ACS
Packaging Poly Bottle
Residue after Ignition 0.0001

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.