missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride (USP/FCC/EP), Fisher Chemical™

Supplier:  Fisher Chemical A68710

 View more versions of this product

Catalog No. A68710

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ammonium Chloride
12125-02-9
5.0
10 kg
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
25517
53.49
EP/FCC/USP
0.5% max. (FCC), 0.5% max. (USP), 0.5% max. (USP/FCC), 1.0% max. (EP), 1.0% max. (EP)
Clear and Colorless (EP)
340°C
White
Solid
ClH4N
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
N.Cl
53.49
CHEBI:31206
99.5 to 100.5% (EP)
Pass Test (EP), Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC/EP)
Poly Pail
1.3mbar at 160°C

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
amine hydrochloride
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
CHEBI:31206
N.Cl
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:0
Instability:1

SDS Documents

Documents

Promotions

Promotions