missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride (USP/FCC/EP), Fisher Chemical™

Catalog No. p-30952
Change view
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
A687500 500 g Poly Bottle
A687212 2.5 kg Poly Bottle
A68710 10 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A687500 Supplier Thermo Fisher Scientific Supplier No. A687500
Only null left

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Melting Point 340°C
Color White
pH 5.0
Physical Form Solid
Quantity 500 g
Formula Weight 53.49
Percent Purity 99.5 to 100.5% (USP)
Grade EP/FCC/USP
Identification Pass Test (EP), Pass Test (FCC), Pass Test (USP), Pass Test (USP/FCC/EP)
Loss on Drying 0.5% max. (FCC), 0.5% max. (USP), 0.5% max. (USP/FCC), 1.0% max. (EP), 1.0% max. (EP)
Packaging Poly Bottle
Appearance Clear and Colorless (EP)
Vapor Pressure 1.3mbar at 160°C
Chemical Name or Material Ammonium Chloride
Show More Show Less
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Harmful if swallowed. May cause skin and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur.

NFPA

Health:2
Flammability:0
Instability:1