missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111020402
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020402 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020402 Supplier Merck Emd Millipore Supplier No. PHR13271G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Ammonium chloride is formed as a by-product during soda manufacturing by Solvay process. It can be synthesized by reacting HCl with aqueous ammonia solutions. Crystals of ammonium chloride (NH4Cl) belong to the cubic crystal system. Featured Industries: Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)

Specifications

CAS 12125-02-9
Vapor Pressure 1 mmHg (160.4°C)
Vapor Density 1.9 (vs. air)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula NH4 Cl
MDL Number MFCD00011420
Melting Point 340°C (sublimation)
Synonym Ammonium chloride; Salmiac
Type Certified Reference Material
Quantity 1 g
Formula Weight 53.49
Grade Certified Reference Material
Show More Show Less