missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, Granular, FCC, 99%, Spectrum™ Chemical
A1167, 12125-02-9, NH4Cl
$533.46 - $7040.14
Chemical Identifiers
| CAS | 12125-02-9 |
|---|---|
| Molecular Formula | ClH4N |
| Molecular Weight (g/mol) | 53.49 |
| MDL Number | MFCD00011420 |
| InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
| IUPAC Name | amine hydrochloride |
| SMILES | N.Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18600583
|
Spectrum Chemical Mfg Cor
A1167500GM |
500 g | Poly Bottle |
Each for $533.46
|
|
||||
|
18600584
|
Spectrum Chemical Mfg Cor
A116725KG |
2.5 kg | Poly Pail |
Each for $1,779.89
|
|
||||
|
18600585
|
Spectrum Chemical Mfg Cor
A116712KG |
12 kg | Poly Pail |
Each for $3,863.64
|
|
||||
|
18600586
|
Spectrum Chemical Mfg Cor
A116745KGBL |
45 kg | Fiber Drum |
Each for $7,040.14
|
|
||||
Description
Spectrum™ Chemical Ammonium Chloride, Granular, FCC is an ingredient in nutritive media for yeasts and many microorganisms. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 12125-02-9 | |
| 53.49 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| N.Cl |
| ClH4N | |
| MFCD00011420 | |
| amine hydrochloride |
Specifications
| 12125-02-9 | |
| 500 g | |
| MFCD00011420 | |
| N.Cl | |
| 53.49 | |
| FCC | |
| Poly Bottle |
| 100% | |
| ClH4N | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| amine hydrochloride | |
| 99% | |
| 0.5% |