missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, Granular, FCC, 99%, Spectrum™ Chemical

A1167, 12125-02-9, NH4Cl

Supplier:  Spectrum Chemical Mfg Cor A1167500GM

 View more versions of this product

Catalog No. 18600583


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Ammonium Chloride, Granular, FCC is an ingredient in nutritive media for yeasts and many microorganisms. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications

Specifications

12125-02-9
500 g
MFCD00011420
N.Cl
53.49
FCC
Poly Bottle
100%
ClH4N
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
99%
0.5%

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
N.Cl
ClH4N
MFCD00011420
amine hydrochloride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions