missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, (Crystalline/ACS Reagent Grade), MP Biomedicals

$164.73 - $521.90

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN19140601
View Documents
MP Biomedicals Inc
0219140601
1 kg
Each for $164.73
Only null left
 
ICN19140605
SDS View Documents
MP Biomedicals Inc
0219140605
5 kg
Each for $521.90
Only null left
 
Description

Description

Specifications

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
25517
amine hydrochloride
ClH4N
MFCD00011420
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
CHEBI:31206
N.Cl

Specifications

12125-02-9
1 kg
MFCD00011420
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
25517
53.5
Ammonium Chloride
Crystalline
ClH4N
ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
N.Cl
53.49
CHEBI:31206
ACS Reagent
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Room Temperature

SDS
missing translation for 'documents'

missing translation for 'documents'