missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, (Crystalline/ACS Reagent Grade), MP Biomedicals
Supplier: MP Biomedicals Inc 0219140601
Specifications
| Ammonium Chloride | |
| Crystalline | |
| ClH4N | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| N.Cl | |
| 53.49 | |
| CHEBI:31206 | |
| ACS Reagent |
| 12125-02-9 | |
| 1 kg | |
| MFCD00011420 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| amine hydrochloride | |
| 25517 | |
| 53.5 |
Chemical Identifiers
| 12125-02-9 | |
| 53.49 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| 25517 | |
| amine hydrochloride |
| ClH4N | |
| MFCD00011420 | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| CHEBI:31206 | |
| N.Cl |
Safety and Handling
Recommended Storage : Room Temperature