missing translation for 'onlineSavingsMsg'
Learn More

Ammonium chloride, ≥99.0%, extra pure, with anti-caking agent

Catalog No. AC396400010
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Bottle
Plastic drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC396400010 1 kg Plastic Bottle
AC396400050 5 kg Plastic drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC396400010 Supplier Thermo Scientific Chemicals Supplier No. 396400010

CAS: 12125-02-9 | ClH4N | 53.49 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
SMILES N.Cl

Specifications

Melting Point 340.0°C
Color White
Physical Form Crystalline Powder
Quantity 1 kg
Assay Percent Range 99.0 to 101% (Argentometry)
Linear Formula NH4Cl
Merck Index 15, 506
Solubility Information Solubility in water: 370g/L (20°C). Other solubilities: soluble in glycerol,slightly soluble in alcohol,practically insoluble in acetone,ether and ethyl,acetate
IUPAC Name azanium;chloride
Formula Weight 53.49
Percent Purity ≥99%
Grade Extra Pure
Packaging Plastic Bottle
Specific Gravity 1.52
Density 1.5200g/mL
Chemical Name or Material Ammonium chloride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wash face,ha
EINECSNumber 235-186-4

RUO – Research Use Only