missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, 5% (w/v), LabChem™

Catalog No. LC110002
Change view
Click to view available options
Quantity:
1 L
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC110002 1 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC110002 Supplier LabChem Supplier No. LC110002

Chemical Identifiers

CAS 12125-02-9 , 7732-18-5
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl

Specifications

Color Colorless
CAS Max % 95, 5
Physical Form Liquid
Quantity 1 L
Linear Formula NH4Cl
Solubility Information Soluble in water
Formula Weight 53.49
Percent Purity 5% (w/v)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1.01g/mL
Decomposition Information Gaseous Ammonia
Density 1.01g/mL
Chemical Name or Material Ammonium Chloride
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature