missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, 2% (w/v), Ricca Chemical
Chemical Identifiers
| CAS | 12125-02-9 , 7732-18-5 |
|---|---|
| Molecular Formula | ClH4N |
| Molecular Weight (g/mol) | 53.489 |
| InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
| Synonym | ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid |
| PubChem CID | 25517 |
| ChEBI | CHEBI:31206 |
| IUPAC Name | azanium;chloride |
| SMILES | [NH4+].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
62416
|
N/A | 500 mL | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
62432
|
N/A | 1 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
6241
|
N/A | 4 L | Natural Poly Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 12125-02-9 , 7732-18-5 | |
| 53.489 | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| CHEBI:31206 | |
| [NH4+].[Cl-] |
| ClH4N | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| 25517 | |
| azanium;chloride |
Specifications
| 12125-02-9 , 7732-18-5 | |
| 100.0, 3.0 | |
| ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid | |
| [NH4+].[Cl-] | |
| 53.489 | |
| CHEBI:31206 | |
| 2% (w/v) |
| 96.0, 1.0 | |
| ClH4N | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| azanium;chloride | |
| 25517 | |
| Aqueous Solution | |
| Ammonium Chloride |