missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Carbonate TS, Ricca Chemical

Catalog No. 622516
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
622516 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 622516 Supplier Ricca Chemical Company Supplier No. 622.516
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material Ammonium Carbonate
Color Colorless
Name Note Test Solution
CAS 506-87-6, 7732-18-5, 1336-21-6
CAS Min % 77.0, 15.41, 5.61
CAS Max % 80.0, 16.04, 5.84
Synonym dihydrogen oxide, dihydrogen monoxide
Grade Laboratory
Packaging Natural Poly Bottle
pH Alkaline
Quantity 500 mL
Physical Form Liquid
Show More Show Less