Learn More
Ammonium bismuth citrate, Bi 48-52%, water ca 2%
CAS: 25530-63-6 | C6H11BiNO7 | 418.134 g/mol
$118.44 - $1225.12
Chemical Identifiers
| CAS | 25530-63-6 |
|---|---|
| Molecular Formula | C6H11BiNO7 |
| Molecular Weight (g/mol) | 418.134 |
| MDL Number | MFCD00036420 |
| InChI Key | JDINTLWFPXWXCT-UHFFFAOYSA-N |
| Synonym | ammonium bismuth citrate |
| PubChem CID | 87101692 |
| IUPAC Name | azane;bismuth;2-hydroxypropane-1,2,3-tricarboxylic acid |
| SMILES | C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Bi] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1700318
|
Thermo Scientific Chemicals
A1700318 |
50 g |
Each for $118.44
|
|
|||||
|
AAA1700330
|
Thermo Scientific Chemicals
A1700330 |
250 g |
Each for $407.78
|
|
|||||
|
AAA170030B
|
Thermo Scientific Chemicals
A170030B |
1000 g |
Each for $1,225.12
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 25530-63-6 | |
| 418.134 | |
| JDINTLWFPXWXCT-UHFFFAOYSA-N | |
| 87101692 | |
| C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Bi] |
| C6H11BiNO7 | |
| MFCD00036420 | |
| ammonium bismuth citrate | |
| azane;bismuth;2-hydroxypropane-1,2,3-tricarboxylic acid |
Specifications
| 25530-63-6 | |
| MFCD00036420 | |
| ammonium bismuth citrate | |
| C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Bi] | |
| 418.134 | |
| 433.12 | |
| Bi 48 to 52%, water ≈2% |
| C6H11BiNO7 | |
| 50 g | |
| JDINTLWFPXWXCT-UHFFFAOYSA-N | |
| azane;bismuth;2-hydroxypropane-1,2,3-tricarboxylic acid | |
| 87101692 | |
| Ammonium bismuth citrate |
Safety and Handling
GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
H302+H312+H332
EINECSNumber : 247-078-4
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only