accessibility menu, dialog, popup

UserName

CAS RN 25530-63-6

IUPAC Name: bismuth(3+) ammonium 2-hydroxypropane-1,2,3-tricarboxylate
Synonyms: bismuth(3+) ammonium citrate
Molecular Weight (g/mol): 416.12
Molecular Formula: C6H9BiNO7
InChi Key: QSBNOZODKXUXSP-UHFFFAOYSA-L
SMILES: [NH4+].[Bi+3].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results