missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bismuth citrate, Bi 48-52%, water ca 2%

Catalog No. AAA1700318
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1700318 50 g
AAA1700330 250 g
AAA170030B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1700318 Supplier Thermo Scientific Chemicals Supplier No. A1700318

CAS: 25530-63-6 | C6H11BiNO7 | 418.134 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 25530-63-6
Molecular Formula C6H11BiNO7
Molecular Weight (g/mol) 418.134
MDL Number MFCD00036420
InChI Key JDINTLWFPXWXCT-UHFFFAOYSA-N
Synonym ammonium bismuth citrate
PubChem CID 87101692
IUPAC Name azane;bismuth;2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES C(C(=O)O)C(CC(=O)O)(C(=O)O)O.N.[Bi]

Specifications

Quantity 50 g
Formula Weight 433.12
Assay Bi 48 to 52%, water ≈2%
Chemical Name or Material Ammonium bismuth citrate
Hazard Category H302+H312+H332
Hazard Statement GHS H Statement
H301-H311-H332
Toxic if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
EINECSNumber 247-078-4
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only