missing translation for 'onlineSavingsMsg'
Learn More

Ammonium bis(oxalato)oxotitanate(IV) hydrate, Puratronic™, 99.998% (metals basis)

CAS: 10580-03-7 | C4H8N2O9Ti | 275.98 g/mol

$157.64 - $547.25

Chemical Identifiers

CAS 10580-03-7
Molecular Formula C4H8N2O9Ti
Molecular Weight (g/mol) 275.98
MDL Number MFCD00149620
InChI Key VBUNWMBOVJSSMH-UHFFFAOYSA-L
Synonym Ammonium titanyloxalate
IUPAC Name diammonium oxotitaniumbis(ylium) dioxalate
SMILES [NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1089509
View Documents
Thermo Scientific Chemicals
01089509
10 g N/A
Only null left
 
AA1089518
View Documents
Thermo Scientific Chemicals
01089518
50 g N/A
Only null left
 
Description

Description

Ammonium bis(oxalato)oxotitanate(IV) hydrate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10580-03-7
275.98
VBUNWMBOVJSSMH-UHFFFAOYSA-L
diammonium oxotitaniumbis(ylium) dioxalate
C4H8N2O9Ti
MFCD00149620
Ammonium titanyloxalate
[NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O

Specifications

10580-03-7
10 g
MFCD00149620
Ammonium titanyloxalate
VBUNWMBOVJSSMH-UHFFFAOYSA-L
diammonium oxotitaniumbis(ylium) dioxalate
276.01 (Anhydrous)
Puratronic
Ammonium bis(oxalato)oxotitanate(IV) hydrate
Crystalline Powder
C4H8N2O9Ti
UN3288
Soluble in water.
[NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
275.98
99.998%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

EINECSNumber : 234-174-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only