CAS RN 10580-03-7
IUPAC Name:
diammonium oxotitaniumbis(ylium) dioxalate
Synonyms:
diammonium oxotitaniumbis(ylium) dioxalate
Molecular Weight (g/mol):
275.98
Molecular Formula:
C4H8N2O9Ti
InChi Key:
VBUNWMBOVJSSMH-UHFFFAOYSA-L
SMILES:
[NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
1
–
3
of
3
results
1
Ammonium titanyl oxalate monohydrate, 98%, pure
CAS: 10580-03-7 Molecular Formula: C4H8N2O9Ti Molecular Weight (g/mol): 275.98 MDL Number: MFCD00149620 InChI Key: VBUNWMBOVJSSMH-UHFFFAOYSA-L Synonym: Ammonium bis(oxalato)oxotitanate(IV) IUPAC Name: diammonium oxotitaniumbis(ylium) dioxalate SMILES: [NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O
2
Ammonium bis(oxalato)oxotitanate(IV) hydrate, Puratronic™, 99.998% (metals basis)
CAS: 10580-03-7 Molecular Formula: C4H8N2O9Ti Molecular Weight (g/mol): 275.98 MDL Number: MFCD00149620 InChI Key: VBUNWMBOVJSSMH-UHFFFAOYSA-L Synonym: Ammonium titanyloxalate IUPAC Name: diammonium oxotitaniumbis(ylium) dioxalate SMILES: [NH4+].[NH4+].O=[Ti++].[O-]C(=O)C([O-])=O.[O-]C(=O)C([O-])=O