missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Bifluoride, Purified, Reagents
Ammonium Bifluoride, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C213050010KG
Specifications
| 240°C | |
| 124.6°C | |
| 1341-49-7 | |
| 10 kg | |
| MFCD00012018 | |
| KVBCYCWRDBDGBG-UHFFFAOYSA-N | |
| amine dihydrofluoride | |
| Pure | |
| 1.5 |
| Ammonium Bifluoride, Purified | |
| Colorless/White | |
| Solid | |
| F2H5N | |
| Ammonium Hydrogen Difluoride, Ammonium Fluoride Hydrofluoride | |
| N.F.F | |
| 57.04 | |
| Poly Pail |
Chemical Identifiers
| 1341-49-7 | |
| 57.04 | |
| KVBCYCWRDBDGBG-UHFFFAOYSA-N | |
| amine dihydrofluoride |
| F2H5N | |
| MFCD00012018 | |
| Ammonium Hydrogen Difluoride, Ammonium Fluoride Hydrofluoride | |
| N.F.F |