missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Bifluoride, Purified, Reagents

Catalog No. p-200021178
Change view
Click to view available options
Quantity:
100 mL
500 g
3 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337506 100 mL
01337503 3 kg
01337502 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01337506 Supplier Labchem Inc Supplier No. C2130500100A6
Only null left

Ammonium Bifluoride, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1341-49-7
Molecular Formula F2H5N
Molecular Weight (g/mol) 57.04
MDL Number MFCD00012018
InChI Key KVBCYCWRDBDGBG-UHFFFAOYSA-N
Synonym Ammonium Hydrogen Difluoride, Ammonium Fluoride Hydrofluoride
IUPAC Name amine dihydrofluoride
SMILES N.F.F

Specifications

Boiling Point 240°C
Melting Point 124.6°C
Color Colorless/White
Physical Form Solid
Quantity 100 mL
Grade Pure
Packaging Poly Bottle
Specific Gravity 1.5
Chemical Name or Material Ammonium Bifluoride, Purified