missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Amidosulfate 98.5+%, TCI America™

Catalog No. p-7135819
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S011725G 25 g
S0117500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S011725G Supplier TCI America Supplier No. S011725G
Only null left

Chemical Identifiers

CAS 7773-06-0
Molecular Formula H6N2O3S
Molecular Weight (g/mol) 114.119
MDL Number MFCD00011429
InChI Key GEHMBYLTCISYNY-UHFFFAOYSA-N
Synonym ammonium sulfamate, ammonium sulphamate, ammate, ikurin, ammonium amidosulfonate, feliderm k, ammate x, sulfamic acid, monoammonium salt, ammonium sulfamidate, ammonium sulphamidate
PubChem CID 24482
ChEBI CHEBI:81950
IUPAC Name azanium;sulfamate
SMILES [NH4+].NS(=O)(=O)[O-]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 25 g
Formula Weight 114.12
Percent Purity ≥98.5% (T)
Chemical Name or Material Ammonium Amidosulfate
EINECSNumber (1)-0404
RTECSNumber WO6125000
TSCA Yes