missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate Buffer, For Iron, Certified, LabChem™

Catalog No. LC109851
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC109851 500 mL
LC109852 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC109851 Supplier LabChem Supplier No. LC109851
Only null left

Specifications

Chemical Name or Material Ammonium Acetate Buffer
Name Note For Iron
CAS 7732-18-5, 64-19-7, 631-61-8
Color Colorless
CAS Max % 70.02, 20.43, 9.55
Decomposition Information Gaseous Ammonia
Physical Form Liquid
Quantity 500 mL
Identification Passes Test
Molecular Formula CH3COONH4
Linear Formula CH3COONH4
Packaging Poly Bottle
Solubility Information Soluble in water
Formula Weight 77.08
Grade Certified
Show More Show Less