missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Acetate, 10% w/v, Reagents

Catalog No. 01336161
Change view
Click to view available options
Quantity: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01336161 Quantity: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336161 Supplier Labchem Inc Supplier No. CA00070020F
Only null left

Ammonium Acetate, 10% w/v, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 631-61-8
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 100°C
Color Colorless
Physical Form Liquid
Quantity 20 L
Percent Purity 10% w/v
Grade USP
Packaging Cubitainer
Specific Gravity 1.02
Chemical Name or Material Ammonium Acetate, 10% w/v