missing translation for 'onlineSavingsMsg'
Learn More

Ammoniacal Buffer, pH 10.0, Ricca Chemical

Catalog No. 616532
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
616532 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 616532 Supplier Ricca Chemical Company Supplier No. 616.532

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

General laboratory reagent used for pH adjustment applications.

Specifications

Chemical Name or Material Ammoniacal Buffer
Name Note pH 10.0
CAS Min % 62.0, 31.76, 4.34
Color Colorless
CAS Max % 64.0, 33.05, 4.51
CAS 1336-21-6, 12125-02-9
Concentration 1X
For Use With (Application) Water Analysis
pH 10
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Natural Poly Bottle
Grade Laboratory
Show More Show Less