missing translation for 'onlineSavingsMsg'
Learn More

Ammoniacal Buffer, pH 10.0, Reagents

Catalog No. 01336043
Change view
Click to view available options
Quantity:
1 L
4 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336043 1 L
01336042 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01336043 Supplier Labchem Inc Supplier No. CA0145001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammoniacal Buffer, pH 10.0, Reagent, use for EDTA titrations. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ammoniacal Buffer, pH 10.0
CAS 7732-18-5, 1336-21-6, 1336-21-6
Boiling Point 36°C
Color Colorless
For Use With (Application) Titration
pH 10
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 0.9964
Sterility Non-sterile
Grade Reagent
Show More Show Less