missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, TS for Dextrose, Reagents

Catalog No. 01336155
Change view
Click to view available options
Quantity: 1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01336155 Quantity: 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01336155 Supplier Labchem Inc Supplier No. CA0018001A
Only null left

Ammonia, TS for Dextrose, Meets USP specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 1336-21-6
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Boiling Point 36°C
Color Colorless
Physical Form Liquid
Quantity 1 L
Grade USP
Packaging Poly Bottle
Specific Gravity 0.96
Chemical Name or Material Ammonia, TS for Dextrose