missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Standard, 100 ppm NH3 (82.2 ppm N), Ricca Chemical

$125.83 - $277.60

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride
SMILES N.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
R6134
View Documents
Ricca Chemical Company
RI6134
120 mL Natural Poly Bottle
Each for $125.83
Only null left
 
613-16
SDS View Documents
Ricca Chemical Company
61316
500 mL Natural Poly Bottle
Each for $240.50
Only null left
 
613-32
View Documents
Ricca Chemical Company
61332
1 L Natural Poly Bottle
Each for $277.60
Only null left
 
Description

Description

Specifications

Chemical Identifiers

12125-02-9
53.49
NLXLAEXVIDQMFP-UHFFFAOYSA-N
CHEBI:31206
N.Cl
ClH4N
MFCD00011420
25517
amine hydrochloride

Specifications

Natural Poly Bottle
98.0,0.03
1 mL = 0.1 mg NH3
ClH4N
Colorless
NLXLAEXVIDQMFP-UHFFFAOYSA-N
amine hydrochloride
120 mL
CHEBI:31206
12125-02-9,7732-18-5
100.0,0.03
Liquid
MFCD00011420
100°C
N.Cl
53.49
25517
Ammonia Standard
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'