missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia solution 25%, Suprapur™, MilliporeSigma™
Supplier: Technidata 1054281000
Specifications
| Colorless | |
| 1336-21-6 | |
| H5NO | |
| 0.903 g/cm3 (20°C) | |
| Soluble (20°C) | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| amine hydrate | |
| 35.05 | |
| CHEBI:18219 | |
| Plastic Bottle | |
| Liquid |
| Suprapur™ | |
| 0.25 | |
| MFCD00066650 | |
| ammonium hydroxide, ammonia water, ammonia aqueous, aquammonia, aqua ammonia, ammonia, aqua, household ammonia, ammonium hydroxide solution, ammonia, monohydrate, ammonium hydroxide nh4 oh | |
| 1 L | |
| N.O | |
| 15.4 to 33.6%(V) | |
| 14923 | |
| Suprapur | |
| 483 hPa (20°C) |
Chemical Identifiers
| 1336-21-6 | |
| 35.05 | |
| VHUUQVKOLVNVRT-UHFFFAOYSA-N | |
| CHEBI:18219 | |
| N.O |
| H5NO | |
| MFCD00066650 | |
| 14923 | |
| amine hydrate |
Safety and Handling
SignalWord: Danger
Recommended Storage : Store below +25°C