missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonia Nitrogen Standard, 10 ppm N (12.2 ppm NH3), Ricca Chemical
$121.69 - $176.66
Chemical Identifiers
| CAS | 12125-02-9 |
|---|---|
| Molecular Formula | ClH4N |
| Molecular Weight (g/mol) | 53.49 |
| MDL Number | MFCD00011420 |
| InChI Key | NLXLAEXVIDQMFP-UHFFFAOYSA-N |
| PubChem CID | 25517 |
| ChEBI | CHEBI:31206 |
| IUPAC Name | amine hydrochloride |
| SMILES | N.Cl |
Chemical Identifiers
| 12125-02-9 | |
| 53.49 | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| CHEBI:31206 | |
| N.Cl |
| ClH4N | |
| MFCD00011420 | |
| 25517 | |
| amine hydrochloride |
Specifications
| Natural Poly Bottle | |
| 98.0,0.0037 | |
| 100.0,0.0039 | |
| Liquid | |
| MFCD00011420 | |
| 100°C | |
| N.Cl | |
| 53.49 | |
| 25517 | |
| Nitrogen Standard |
| 12125-02-9,7732-18-5 | |
| 10ppm N as Ammonia (12.2ppm NH3) | |
| 1 mL = 0.01 mg N | |
| ClH4N | |
| Colorless | |
| NLXLAEXVIDQMFP-UHFFFAOYSA-N | |
| amine hydrochloride | |
| 120 mL | |
| CHEBI:31206 |