missing translation for 'onlineSavingsMsg'
Learn More

Precision Laboratories, Inc Ammonia Leak Detection Cloth

Catalog No. 14853276
Change view
Click to view available options
Dimensions (L x W):
40 x 40 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
14853276 40 x 40 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14853276 Supplier Precision Laboratories, Inc Supplier No. 50514040

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonia leak detection cloth detects leaks in sealed incubators and industrial systems, including containers and filtration handling ammonia gas

  • The Ammonia Leak Detection Cloth is reversible.
  • As long as the cloth reverts back to the original goldenrod yellow color, it will continue to change when exposed to ammonia vapors.
  • However, we aren't certain how many times it can be reused (it is somewhat dependent on the use).
  • Eventually, it will not revert back to its original color and can no longer be reused. It works best if exposed to vapors.
  • If the cloth gets wet, it will not be reusable.
  • The cloth is sensitive to vapors from a 1% solution of Ammonia.
  • Ammonia Leak Detection Cloth
    • 1. Place the Ammonia Leak Detection Cloth near a suspected leak.
    • 2. The cloth will turn blue immediately upon detection.

Specifications

Color Goldenrod
Dimensions (L x W) 40 x 40 in.
Type Vapor Leak Detection Cloth
For Use With (Application) Color changes to blue upon ammonia detection