missing translation for 'onlineSavingsMsg'
Learn More

Ammonia ISE Fill Solution, Ricca Chemical

Catalog No. p-200004035
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01213120 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01213120 Supplier Ricca Chemical Company Supplier No. R3161200500A
Only null left

Specifications

Packaging 500 mL HDPE Bottle
Certificate of Analysis Yes
CAS Max % 0.07%, 99.38%, 0.55%
CAS 12125-02-9, 107-21-1, 7732-18-5
CAS Min % 99.38%, 0.07%, 0.55%
Melting Point 0.0°C
Boiling Point 100°C
Quantity 500 mL
ShelfLife 12 Months