missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Buffer, for Hardness Testing, Ricca Chemical

Catalog No. R6161
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R6161 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R6161 Supplier Ricca Chemical Company Supplier No. 6161

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Hardness Testing Buffer

Specifications

Chemical Name or Material Ammonia Buffer, for Hardness Testing
Melting Point ∼0°C
CAS 7732-18-5, 12125-02-9, 1336-21-6, 12135-76-1
Boiling Point ∼100°C
CAS Min % 84.9, 7.44, 7.14, 0.51
Color Green-Yellow
CAS Max % 84.9, 7.44, 7.14, 0.51
Density 0.9
Concentration 1X
For Use With (Application) Water Analysis
pH 9.90 to 10.10
Physical Form Liquid
Quantity 4 L
Solution Type Buffer
Packaging Natural Poly Bottle
Solubility Information Miscible in water
Grade Laboratory
Show More Show Less