missing translation for 'onlineSavingsMsg'
Learn More

Ammonia-Ammonium Chloride Buffer TS, Reagents

Catalog No. 01336048
Change view
Click to view available options
Quantity:
500 mL
1 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01336048 1 L
01336047 20 L
01336046 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01336048 Supplier Labchem Inc Supplier No. CA0143001A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ammonia-Ammonium Chloride Buffer TS, Meets USP specifications, use for EDTA titrations. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Ammonia-Ammonium Chloride Buffer TS
CAS 7732-18-5, 1336-21-6, 1336-21-6
Boiling Point 36°C
Color Colorless
For Use With (Application) Titration
pH 10
Physical Form Liquid
Quantity 1 L
Solution Type Buffer
Packaging Poly Bottle
Specific Gravity 0.9
Sterility Non-sterile
Grade USP
Show More Show Less