missing translation for 'onlineSavingsMsg'
Learn More

Amlodipine besylate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111012471
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012471 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012471 Supplier Merck Emd Millipore Supplier No. PHR11851G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Amlodipine besylate is a calcium channel blocker, which inhibits the trans membrane influx of calcium ions into vascular smooth muscles and cardiac muscle. It belongs to the dihydropyridine family. It is used in combination with atorvastatin calcium to treat vasospastic angina, chronic stable angina, hypertension, in elevated serum triglyceride levels, primary dysbetalipoproteinemia. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 111470-99-6
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C20H25 ClN2O5 · C6H5 SO3 H
MDL Number MFCD00887594
Synonym 2-[(2-Aminoethoxy)-methyl]-4-(2-chlorophenyl)-1,4-dihydro-6-methyl-3,5- pyridinedicarboxylic acid 3-ethyl 5-methyl ester benzene sulfonate; Norvasc
Type Certified Reference Material
Quantity 1 g
Formula Weight 567.05
Grade Certified Reference Material