missing translation for 'onlineSavingsMsg'
Learn More

Amlexanox 98.0+%, TCI America™

Catalog No. p-7117883
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24011G 1 g
A24015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24011G Supplier TCI America Supplier No. A24011G
Only null left

Chemical Identifiers

CAS 68302-57-8
Molecular Formula C16H14N2O4
Molecular Weight (g/mol) 298.30
MDL Number MFCD00864790
InChI Key SGRYPYWGNKJSDL-UHFFFAOYSA-N
Synonym amlexanox, aphthasol, amoxanox, 2-amino-7-isopropyl-5-oxo-5h-chromeno 2,3-b pyridine-3-carboxylic acid, amlenanox, solfa, amlexanoxo, amlexanoxum, amlexanoxum latin, amlexanoxo spanish
PubChem CID 2161
ChEBI CHEBI:31205
IUPAC Name 2-amino-5-oxo-7-(propan-2-yl)-5H-chromeno[2,3-b]pyridine-3-carboxylic acid
SMILES CC(C)C1=CC=C2OC3=NC(N)=C(C=C3C(=O)C2=C1)C(O)=O

Specifications

Melting Point 300°C
Color White-Yellow
Quantity 1 g
Formula Weight 298.30
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Amlexanox
RTECSNumber DJ3102000
TSCA No