missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline N-β-D-glucuronide Solution, 100 μg/mL in Acetonitrile: water (1:1), Certified Reference Material, Ampule of 1 mL, Cerilliant™

Catalog No. A1281ML
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1281ML 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A1281ML Supplier MilliporeSigma Supplier No. A1281ML
  • A certified solution standard of Amitriptyline N-β-D-glucuronide, a metabolite of the tricyclic antidepressant amitriptyline.
  • Sold under trade names such as Amitrip, Elavil, and Levate, amitriptyline is prescribed to treat a number of mental disorders ranging from major depressive disorder and anxiety disorder to bipolar disorder.
  • This solution-based Certified Reference Material is applicable for use in LC/MS or GC/MS testing applications in clinical toxicology, forensic analysis, workplace drug testing, and pharmaceutical research.

Specifications

Chemical Name or Material Amitriptyline N-beta-D-glucuronide
Packaging Ampule
CAS 112806-33-4
Molecular Formula C26H31NO6
Concentration Acetonitrile: Water (1:1), 100 μg/mL
For Use With (Equipment) Liquid Chromatography (LC); Gas Chromatography (GC)
Solution Type Standard Solution
Type Certified Reference Material
Industry Type Clinical Testing
Molecular Weight (g/mol) 453.53
Quantity 1 mL
Grade Certified
Show More Show Less