missing translation for 'onlineSavingsMsg'
Learn More

Amitriptyline Hydrochloride 98.0+%, TCI America™

Catalog No. A090825G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A090825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A090825G Supplier TCI America Supplier No. A090825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 549-18-8
Molecular Formula C20H24ClN
Molecular Weight (g/mol) 313.869
MDL Number MFCD00012537
InChI Key KFYRPLNVJVHZGT-UHFFFAOYSA-N
Synonym amitriptyline hydrochloride, amitriptyline hcl, annoyltin, tryptizol, domical, elavil hydrochloride, lentizol, saroten, amavil, ami-anelun
PubChem CID 11065
IUPAC Name 3-(5,6-dihydrodibenzo[2,1-b
SMILES CN(C)CCC=C1C2=CC=CC=C2CCC3=CC=CC=C31.Cl

Specifications

Melting Point 195°C
Color White
Quantity 25 g
UN Number 2811
Formula Weight 313.87
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Amitriptyline Hydrochloride
RTECSNumber HO9450000
TSCA Yes